Learn More
Epredia™ Papanicolaou OG-6 and Modified OG Stains
Stain gynecological specimens with Epredia™ Papanicolaou Stain OGs.
Brand:  Epredia™ 75211
79.38 GBP valid until 2024-11-29
Use promo code "21706" to get your promotional price.
 View more versions of this product
Product Code. 10444340
  • £121.00 / 3.79 L
Explore more special offers
Description
Description
  • OG-6 stains the keratinized areas of the cell
  • Modified OG is a Gill modification that minimizes precipitation and reduces staining time
  • Use OG-6 with EA-36 or EA-50 when staining gynecological specimens
  • Use modified OG with EA-65 for staining non-gynecological specimens
Specifications
Specifications
OG-6 Papanicolaou Stain
1 gal.