Learn More
Paper Mercuric Bromide, European Pharmacopoeia Grade, Fisher Chemical™ Product Code.: 12960454
compliance-icons compliance-icons compliance-icons
Paper Mercuric Bromide, European Pharmacopoeia Grade, Fisher Chemical™
Product Code. 12960454
50 mL
Click to view available options
Quantity:
50 mL
Product Code. 12960454
Brand: Fisher Chemical RP 118
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 50 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1006563 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 118
Lot Number1029191 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 118
Lot Number1023648 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberRP 118
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercuric Bromide Paper. European Pharmacopoeia Ref: 1052101 (RP 118)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P314-Get medical advice/attention if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .
Product Content Correction
Product Title
Paper Mercuric Bromide, European Pharmacopoeia Grade, Fisher Chemical™ >