Learn More
Paper Starch iodate EU Pharmacopoeia, Fisher Chemical™
Product Code. 12940544
50 mL
Click to view available options
Quantity:
50 mL
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 50 mL
Product Content Correction
Product Title
Paper Starch iodate EU Pharmacopoeia, Fisher Chemical™ >