missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPOLB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£122.57 - £303.85
Specifications
Antigen | PAPOLB |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890734
|
Proteintech
12821-1-AP-20UL |
20 μL |
£122.57
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880734
|
Proteintech
12821-1-AP-150UL |
150 μL |
£303.85
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Testis-specific poly(A) polymerase.Specifications
PAPOLB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q9NRJ5, Q9WVP6 | |
304300, 56522, 56903 | |
PAPOLB Fusion Protein Ag3487 | |
Primary | |
-20°C | |
PAPOLB |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
PAP beta, PAPOLB, PAPT, Poly(A) polymerase beta, TPAP | |
Papolb | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |