Learn More
PAPPA Mouse anti-Human, Unconjugated, Clone: 1G5A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66962-1-IG-150UL
 View more versions of this product
Product Code. 16819295
  • £349.00 / 150µL
Explore more special offers
Description
Description
The PAPPA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PAPPA. This antibody recognizes human antigen. The PAPPA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PAPPA
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1G5A7
Unconjugated
150 μL
5069
IgG1