missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paraffin Liquid, Technical, d=0.88, for Oil Baths, Fisher Chemical™
£94.25 - £534.00
Chemical Identifiers
CAS | 8042-47-5 |
---|---|
Molecular Formula | MFCD00131611 |
Molecular Weight (g/mol) | 0.00 |
MDL Number | MFCD00131611 |
InChI Key | FFNDMZIBVDSQFI-UHFFFAOYSA-N |
Synonym | delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride |
PubChem CID | 68245 |
ChEBI | CHEBI:38701 |
IUPAC Name | 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride |
SMILES | * |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10744571
|
Fisher Chemical
P/0320/17 |
2.5 L | Amber glass bottle |
£94.25
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10579380
|
Fisher Chemical
P/0320/25 |
25 L | Plastic container |
£534.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Paraffin Liquid, Technical, d=0.88, for Oil Baths
Chemical Identifiers
8042-47-5 | |
0.00 | |
FFNDMZIBVDSQFI-UHFFFAOYSA-N | |
68245 | |
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride |
MFCD00131611 | |
MFCD00131611 | |
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride | |
CHEBI:38701 | |
* |
Specifications
8042-47-5 | |
310°C | |
MFCD00131611 | |
FFNDMZIBVDSQFI-UHFFFAOYSA-N | |
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride | |
68245 | |
CHEBI:38701 | |
Viscous Liquid |
-9°C | |
MFCD00131611 | |
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride | |
* | |
0.00 | |
>2 at 101kPa | |
>19 St (40°C) |