missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Paraffin Type 1, 3, 6, 9

Find the perfect fit with four different blends of highly refined paraffin and polymers, offering a choice of infiltration times and cutting characteristics.

Brand:  Epredia™ 8336

Code : A0

Additional Details : Weight : 9.80000kg


 View more versions of this product

Product Code. 12066669

  • £110.00 / Pack of 10
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

  • All paraffins melt between 55° and 57°C and are triple filtered to 0.5μm
  • Flake form has a greater surface-to-weight ratio than pellets, resulting in a faster melting time
  • Type 1 contains no polymer and offers rapid infiltration
  • Type 3 contains minimal polymer and offers fast infiltration
  • Type 6 is harder than Type 3 and allows for thinner sections
  • Type 9 has the highest polymer content available and yields extremely thin sections

Ordering Information:
Packaged in convenient, easy-to-open, resealable bags

Specifications

Specifications

54° to 57°C
Paraffin
2 lb.
Infiltration and Embedding
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H335-May cause respiratory irritation.
  • H371-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P314-Get medical advice/attention if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : parafin waxes and hydrocarbon waxes, 2,6-di-tert-butyl-p-cresol, microcrystalinewax

SDS
Documents

Documents

Certificates
Special Offers

Special Offers