Learn More
Cole-Parmer™ WD-200 Series Wax Dispenser
Prevent wax blockage with heated tap with the WD-200 Series Wax Dispenser.
Brand:  Cole-Parmer™ 85230-07
Product Code. 15804285
  • £2845.00 / Each
Explore more special offers
Description
Description
  • Accurate temperature control with digital display
  • Constructed of stainless steel with antimicrobial coating to inhibit bacteria growth
  • Enhanced safety with high temperature cut-off at 120°C
  • Fully insulated tank prevents heat loss and ensures the outer surface stays cool
This paraffin wax dispenser provides ultrafast heating for on-demand molten wax, ideal for your histology and pathology applications. The heated tap prevents blockage providing a smooth even flow of wax every time. The nondrip lever has three settings—off, on-demand pull dispense, and continuous flow. The 0.5 mm mesh filter screen prevents coarse particles from entering the tap.
Temperature can be set from ambient to 70°C using digital temperature control panel. The on/off temperature control keeps accuracy to ±1°C. The 7.5 L capacity tank can melt up to 13.2 lb (6 kg) of pelletized wax at a time. When set at 65°C, the unit will take approximately 90 minutes to fully melt 11 lb. (5 kg) of pelletized wax.
Specifications
Specifications
455 mm
220/240 V
7.5 L
MH8524
50/60 Hz
2 gal.
Paraffin Wax Dispenser