missing translation for 'onlineSavingsMsg'
Learn More

Paraffin wax, pure, granular, ACROS Organics™

Product Code. 10711452
Click to view available options
Quantity:
2 kg
10 kg

Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-74-2
Molecular Formula CnH2n+2
Molecular Weight (g/mol) 341.451
MDL Number MFCD00132833
InChI Key JWHAUXFOSRPERK-UHFFFAOYSA-N
Synonym propafenone, rythmol, propafenona, propafenonum, propafenonum inn-latin, propafenona inn-spanish, propafenone inn:ban, gnf-pf-4594, propafenone-hcl, propafenone inn
PubChem CID 4932
ChEBI CHEBI:63619
IUPAC Name 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one
SMILES CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O

Specifications

Melting Point 58.0°C to 62.0°C
Color White
Boiling Point 322.0°C
Flash Point 198°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 2 kg
Merck Index 14,7023
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in alcohol (soluble in warm alcohol),insoluble in acids,soluble in benzene,chloroform,ether,carbon,disulfide,ligroin,turpentine,miscible when melted with wax,fats
Viscosity 3-4.5 mPa.s (98.9°C)
Grade Pure
Physical Form Pellets
Chemical Name or Material Paraffin wax
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'