Learn More
Paraffine Solution, Fluka
Brand: Honeywell Fluka 34664-2.5L
821.75 GBP valid until 2024-12-31
Use promo code "23289" to get your promotional price.
Additional Details : Weight : 0.78000kg Transport : UN number : 1145 Chem class : 3 Pack group : II
Specifications
Paraffine Solution | |
79°C | |
0.78 g/cm3 | |
PANPEHA Indicator Paper | |
2.5 L | |
-18°C | |
UN1145 |
110-82-7,8002-74-2 | |
Colorless | |
<10% | |
Liquid | |
Mixture | |
Plastic Bottle |
Safety and Handling
- Paraffinlösung Nr.9 für PANPEHA-Indikatorpapier
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : cyclohexane