missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Paraformaldehyde, 10% w/v aq. soln., methanol free Product Code.: 17876953

Thermo Scientific Chemicals Paraformaldehyde, 10% w/v aq. soln., methanol free

Product Code. 17876953
10 x 10 mL
Click to view available options
Quantity:
10 x 10 mL
This item is not returnable. View return policy

Product Code. 17876953

Brand: Thermo Scientific Chemicals 047317.9M

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solution is micro-filtered and supplied in ampules.

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Linear Formula (CH2O)n
Quantity 10 x 10 mL
Beilstein 102769
Merck Index 14,7025
Solubility Information Soluble in chlorophenol and many organic solvents. Insoluble in ether and alcohol.
Concentration or Composition (by Analyte or Components) 10% w/v aq. soln.
Physical Form Liquid
Chemical Name or Material Paraformaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY15K049 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number047317.9M
Lot NumberP26K016 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number047317.9M
Lot NumberW03H002 Certificate TypeCertificate of Analysis Date08/02/2023 Catalog Number047317.9M
Lot NumberU05H024 Certificate TypeCertificate of Analysis Date08/02/2023 Catalog Number047317.9M
Lot NumberU17E093 Certificate TypeCertificate of Analysis Date08/02/2023 Catalog Number047317.9M

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Paraformaldehyde, 10% w/v aq. soln., methanol free >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.