missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Paraformaldehyde, 16% w/v aq. soln., methanol free Product Code.: 11490570

Thermo Scientific Chemicals Paraformaldehyde, 16% w/v aq. soln., methanol free

Product Code. 11490570
2 x 10 mL
Click to view available options
Quantity:
2 x 10 mL
10 x 10 mL
This item is not returnable. View return policy

Product Code. 11490570

Brand: Thermo Scientific Chemicals 043368.9L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paraformaldehyde, CH2O, CAS Number-30525-89-4, methylene oxide, oxomethane, paraform, oxymethylene, formalin, formol, methyl aldehyde, paraformaldehyde, formic aldehyde, methanal, 10 x 10mL, CHEBI:16842, Hazard Class: 9; Packaging Group: III, 200-001-8, H312+H332-H315-H317-H319-H335-H341-H350

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Packaging Ampule
MDL Number MFCD00133991
Quantity 2 x 10 mL
Merck Index 14,7025
Solubility Information Soluble in chlorophenol and many organic solvents. Insoluble in ether and alcohol.
Physical Form Liquid
Chemical Name or Material Paraformaldehyde, Methanol free

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT30B038 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog Number043368.9L,043368.9M
Lot NumberY14K041 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number043368.9L,043368.9M
Lot NumberT15K021 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number043368.9L,043368.9M
Lot NumberR23K019 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number043368.9L,043368.9M
Lot NumberN16K030 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number043368.9L,043368.9M
compliance-icons compliance-icons
Product Identifier
  • Paraformaldehyde, 16% w/v aqueous solution
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P363-Wash contaminated clothing before reuse.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Paraformaldehyde
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Paraformaldehyde, 16% w/v aq. soln., methanol free >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.