missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS Product Code.: 15424389

Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS

Product Code. 15424389
250 mL
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15424389

Brand: Thermo Scientific Chemicals J61899.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paraformaldehyde, CH2O, CAS Number-30525-89-4, methylene oxide, paraformaldehyde, formalin, paraform, methyl aldehyde, formic aldehyde, formol, oxymethylene, methanal, oxomethane, 500mL, CHEBI:16842, H317-H318-H351

Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with brass, steel, copper, acid anhydrides, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Quantity 250 mL
Solubility Information Miscible with water.
Chemical Name or Material Paraformaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP28L500 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog NumberJ61899.AK,J61899.AP
Lot NumberP15L509 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog NumberJ61899.AK,J61899.AP
Lot NumberQ11L500 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog NumberJ61899.AK,J61899.AP
Lot NumberP15L510 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberJ61899.AK,J61899.AP
Lot NumberN25L500 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberJ61899.AK,J61899.AP
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paraformaldehyde, 4% in PBS
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : paraformaldehyde, sodium chloride, phosphoric acid, potassium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.