missing translation for 'onlineSavingsMsg'
Learn More

Paraquat monopyridone iodide, Honeywell Fluka™

Product Code. 15961131
Click to view available options
Quantity:
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

CAS 35022-71-0
Packaging Glass Bottle
Quantity 25 mg
Grade Analytical Standard
For Use With (Application) Herbicides
Physical Form Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'