missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline base, Thermo Scientific Chemicals

Product Code. 10703085
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 467-62-9
Molecular Formula C19H19N3O
Molecular Weight (g/mol) 305.38
MDL Number MFCD00036222
InChI Key KRVRUAYUNOQMOV-UHFFFAOYSA-N
Synonym pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol
PubChem CID 10084
IUPAC Name tris(4-aminophenyl)methanol
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N

Specifications

Melting Point 205.0°C
Color Purple
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 50 g
Beilstein 13, 733
Solubility Information Solubility in water: very slightly soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 305.38
Physical Form Crystalline Powder
Chemical Name or Material Pararosaniline base
compliance-icons
Product Identifier
  • Pararosaniline base
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'