missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pararosaniline base, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 205.0°C |
Color | Purple |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 10 g |
Beilstein | 13, 733 |
Solubility Information | Solubility in water: very slightly soluble. Other solubilities: soluble in alcohol,slightly soluble in ether |
Formula Weight | 305.38 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Pararosaniline base |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Pararosaniline base
- Warning
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'