missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARD3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PARD3 |
---|---|
Concentration | 300 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887923
|
Proteintech
11085-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877923
|
Proteintech
11085-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PARD3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PARD3. This antibody recognizes human, mouse, rat antigen. The PARD3 antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Specifications
PARD3 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
300 μg/mL | |
Polyclonal | |
Rabbit | |
56288 | |
Primary |