Learn More
Paricalcitol-d6, MedChemExpress
MedChemExpress Paricalcitol-d6 is the deuterium labeled Paricalcitol. Paricalcitol is a drug used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure.
Brand:  Medchem Express HY-76585
Encompass_Preferred
 View more versions of this product
Product Code. 16470648
  • £3296.04 / 5mg
Explore more special offers
Description
Description
Paricalcitol-d6 is the deuterium labeled Paricalcitol. Paricalcitol is a drug used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure.
Specifications
Specifications
Paricalcitol-d6
5 mg
Research
DMSO : 100 mg/mL (236.59 mM; Need ultrasonic)
422.67
99.64%
Metabolism-sugar/lipid metabolism
2070009-67-3
White
C27H38D6O3
C[C@]([C@@H]([C@@H](/C=C/[C@@H](C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)C)CC1)(CCC/2)[C@@]1(C2=C\C=C3C[C@H](C[C@@H](C/3)O)O)[H]
422.67
Research
Solid
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
Research purposes only