missing translation for 'onlineSavingsMsg'
Learn More

Checkers Park-It™, Black/White

The rubber Park-It™ Parking Curb is used to effectively guide cars when pulling into a parking place. It is profiled at 10 cm high, safeguarding potential damage to today’s low, front-end car designs. They are more resistant than concrete or plastic curbs.

Brand:  Checkers GNRS1420WB

Additional Details : Weight : 10.50000kg

 View more versions of this product

Product Code. 17732318

  • £45.80 / Each
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

  • The Park-It™ parking curb is constructed of 100% recycled rubber and is used to effectively guide cars when pulling into a parking stall
  • It is profiled at 10 cm high, safeguarding potential damage to today's low, front-end car designs
  • Unlike concrete or plastic, which necessitate frequent replacements, the Park-It™'s recycled rubber construction makes it safer and more durable
  • In-moulded highway reflective tape (available in yellow or white) ensures visibility and pedestrian safety
  • The Park It™ comes with pre-moulded holes and can be shipped with rebar spikes or lag bolts for installation
  • They are lightweight (16 kg for an 18 cm Park-It™) and flexible; conforming to the contour of uneven road surfaces
  • Park-It™ parking curbs are available in 90 cm, 120 cm and 180 cm lengths and are covered by a 3-year warranty; 1 year on the tape (under normal use)
Specifications

Specifications

47 in.
Parking Curb
Recycled Rubber
Black, White
15 cm
120 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers