Learn More
Cole-Parmer™ Parker Hannifin™ P-P-6 Inserts for Soft Tubing
Inserts fit in the end of the soft tubing to provide the rigidity required for adequate compression and a proper seal
Brand: Cole-Parmer™ 06391-30
Code : A0
Additional Details : Weight : 0.02000kg
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Compression fittings are an excellent choice for connecting rigid tubing systems
- Very good chemical resistance
Specifications
Inserts for Soft Tubing | |
Not Autoclavable | |
1/4 in. | |
3/8 in. | |
Ethylene Oxide |
Compression Connection | |
Polypropylene | |
1/4 in. | |
1/4 in. | |
White |