Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Union Tee
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Brand:  Cole-Parmer™ 31320-29
 View more versions of this product
Product Code. 11745738
  • £61.50 / Each
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Superior chemical resistance with durable PFA composition.
  • Get a tight seal at higher pressures.
  • Withstand higher pressures than barbed fittings.
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Specifications
Compression Union Tee
PFA
1/8 in.
Autoclavable
Compression Union Tee
Autoclave or Ethylene Oxide