Learn More
3M™ Particulate Filter, 2000 Series
Product Code. 12322329
Click to view available options
:
Filter type: P2R
Product Code. 12322329
Brand: 3M™ 7100015050
Only null left
Add to basket
Add to basket
Offers low breathing resistance. The 3M Particulate Filter uses bayonet-style cartridges for half- or full-facemasks.
  • Lightweight particulate filters with low breathing resistance
  • Bayonet-style cartridge connection for fitting to facepiece
  • Filters approved to EN143:2000
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN143 : 2000, CE Approved
Connection Type Bayonet
Description Solid and liquid particles
Filter Type P2R
Product Type Particulate Filter
For Use With (Equipment) Half- and Full-face masks