missing translation for 'onlineSavingsMsg'
Learn More

Parvalbumin Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 29312-1-AP-20UL

 View more versions of this product

Product Code. 17669656

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Parvalbumin Polyclonal antibody apecifically detetects Parvalbumin antigen in Human, Mouse Samples. It is validated for Immunohistochemistry, ELISA.
Specifications

Specifications

Parvalbumin
Polyclonal
Unconjugated
NM_002854
Rabbit
Antigen affinity purification
Primary
Human, Mouse
Liquid
Immunohistochemistry, ELISA
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
PVALB
Parvalbumin fusion protein Ag30189
20 μL
5816
Store at -20°C. Stable for one year after shipment.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.