Learn More
Novacyt - Microgen Bioproducts Path-Chek Protein Swab Test
Simple and instant swab test for the assessment of cleaning efficiency by the detection of protein and possible allergens.
Brand:  Novacyt - Microgen Bioproducts PC006
Product Code. 15907132
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Path-Chek Hygiene Test
  • Rapid test system designed for the detection of a range of bacteria from work surfaces and the processing environment.
  • Simple and instant swab test for the assessment of cleaning efficiency by the detection of protein and possible allergens.
Specifications
Specifications
Protein, possible allergens
Swab Test
100 Tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Novacyt - Microgen Bioproducts Path-Chek Protein Swab Test > Detects: Protein, possible allergens, 100 tests