missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Path-ID™ qPCR Master Mix
Ambion™ Path-ID™ qPCR Master Mix is a convenient enzyme, buffer, and dNTP mix for TaqMan™ probe-based, real-time PCR detection of DNA targets.
£151.00 - £582.00
Specifications
Content And Storage | 2X qPCR Master Mix is stored at –20°C. Nuclease-free water may be stored at any temperature. |
---|---|
Detection Method | Primer-probe |
Form | Liquid |
Format | Tube |
PCR Method | qPCR |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
30057778
|
Applied Biosystems™
4388644 |
500 Reactions |
£582.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30057772
|
Applied Biosystems™
4388643 |
100 Reactions |
£151.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ambion™ Path-ID™ qPCR Master Mix is optimized for targets of importance in animal health and to function in the presence of PCR inhibitors. The mix is supplied in one tube suitable for 100 reactions. With the exception of PCR primers and probes for your target, the master mix contains everything you need for real-time PCR, including:
- AmpliTaq Gold™ ultrapure thermostable DNA polymerase designed for automatic hot start, to minimize nonspecific PCR products
- Optimized buffer and dNTP concentration, for reliable PCR
- Passive internal reference, based on proprietary ROX™ dye, for precise data analysis.
Specifications
2X qPCR Master Mix is stored at –20°C. Nuclease-free water may be stored at any temperature. | |
Liquid | |
qPCR | |
Fast or Standard | |
Ambion™, Path-ID™ | |
Dry Ice | |
Built-In Hot Start |
Primer-probe | |
Tube | |
AmpliTaq Gold DNA Polymerase | |
ROX (Pre-mixed) | |
UP (Ultra Pure) | |
Animal Health, Pathogen Detection | |
DNA (Genomic), cDNA |
For Veterinary Use Only.