missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts PathFlow Rotavirus/Adenovirus Combi Test Kit
Microgen Bioproducts PathFlow Rotavirus / Adenovirus combi Test Kit
Brand: Novacyt - Microgen Bioproducts M595CE
This item is not returnable.
View return policy
Description
- Immunoassay for the qualitative detection of adenovirus
Specifications
ISO13485 ISO9001 | |
Yes | |
Rotavirus and Adenovirus Antibodies | |
Swab | |
Test Strip | |
Specimen Collection/ Stool Container, Timer, Centrifuge and Pipette | |
Plastic | |
Animal | |
25 Tests and 1 Bottle (3 mL) | |
Rotavirus and adenovirus | |
Eukaryota |
2°C to 30°C | |
Non-waived | |
25 Tests | |
Rotavirus: 97.3% Adenovirus: 95.2% | |
A Qualitative, Lateralflow Immunoassay for The Detection of Rotavirus and Adenovirus in Human Faeces Specimen | |
25 Tests | |
1/Test | |
Carton with sealed foil pouches | |
Contains anti-rotavirus antibody and anti-adenovirus antibody coated particles and anti-rotavirus antibody and anti-adenovirus antibody coated on the membrane | |
Rotavirus: 97.1% Adenovirus: 97.7% | |
15 minutes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novacyt - Microgen Bioproducts PathFlow Rotavirus/Adenovirus Combi Test Kit > PathFlow Rotavirus/Adenovirus Combi Test Kit
Spot an opportunity for improvement?Share a Content Correction