Learn More
Intelsius™ PathoPouch™ 95kPa Xcel
The PathoPouch™ 95kPa Xcel is an extra-large flexible secondary pouch designed to safely transport samples and withstand 95kPa of pressure differential in compliance with category A and B UN transport regulations. Supplied with large six-bay to keep up to six samples safe and secure.
Brand: Intelsius™ BP008
Description
Suitable for the transportation of a variety of samples including:- Diagnostic Samples
- Category A and B Infectious Substances
- Blood Vials
- Swabs
Specifications
6 Bays | |
Leak-proof Sample Pouch | |
460 x 334 mm (18.1 x 13.1 in.) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.