Learn More
Intelsius™ PathoSorb Absorbent Sheet
The PathoSorb Absorbent Sheet is a lint free absorbent sheet that reduces the contamination risk and satisfies category A and B transport regulations when transporting samples.
Brand:  Intelsius™ AB007
Encompass_Preferred
Product Code. 17318589
  • £305.98 / Pack of 200
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 600 mL saline absorbency capacity
  • 693 mL water absorbency capacity
  • Lint free
  • Super Absorbent
  • Suitable in supporting UN3373 Category B and UN2900/UN2814 Category A transport regulations 
Specifications
Specifications
Absorbent Sample Sheet
470 x 295 mm (18.5 x 11.6 in.)
600 mL