missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Patrick Safety Shoes
An ideal choice when optimal foot protection is required. A strong, solid steel plate in the sole and resilient leather uppers defend the feet from unexpected hazards. Perfectly balanced, anatomical arch support will keep feet feeling fit and fresh.
£147.00 - £164.00
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | Black |
Design | Patrick |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15998931
|
Emma Safety Footwear
MM73084637 |
37 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15908941
|
Emma Safety Footwear
MM73084638 |
38 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15918941
|
Emma Safety Footwear
MM73084639 |
39 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15928941
|
Emma Safety Footwear
MM73084640 |
40 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15938941
|
Emma Safety Footwear
MM73084641 |
41 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15948941
|
Emma Safety Footwear
MM73084642 |
42 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15958941
|
Emma Safety Footwear
MM73084643 |
43 |
£164.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15968941
|
Emma Safety Footwear
MM73084644 |
44 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15978941
|
Emma Safety Footwear
MM73084645 |
45 |
£164.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15988941
|
Emma Safety Footwear
MM73084646 |
46 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15998941
|
Emma Safety Footwear
MM73084647 |
47 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15908951
|
Emma Safety Footwear
MM73084648 |
48 |
£147.00
1 pair |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Laces | |
Patrick | |
Leather |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title