missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20570-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to PAX7. It will not cross react with other PAX members.
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene.Specifications
PAX7 | |
Polyclonal | |
Unconjugated | |
PAX7 | |
HUP1, paired box 7, Paired box protein Pax 7, PAX7, PAX7B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18509, 5081 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P23759, P47239 | |
PAX7 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PAX7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction