missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX8 Mouse, Clone: PAX8/1491, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7849-MSM1-P0
This item is not returnable.
View return policy
Specifications
PAX8 | |
Nuclear | |
PAX8/1491 | |
Unconjugated | |
XP_016860367 | |
Mouse | |
62kDa | |
Protein A/G Purified | |
RUO | |
7849 | |
IgG2b |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
PAX8 | |
Recombinant human PAX8 fragment (aa60-261) | |
Raji cells. Renal Cell Carcinoma (RCC) | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |