missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Dulbecco's Phosphate Buffered Saline (10X) without Calcium and Magnesium
Stock solution used to prepare 1X D-PBS in cell culture grade water.
Brand: MP Biomedicals™ 091960454
39.12 GBP valid until 2024-12-31
Use promo code "21951" to get your promotional price.
This item is not returnable.
View return policy
Description
Dulbecco's Phosphate Buffered Saline 10X is a stock solution used to prepare 1X D-PBS in cell culture grade water. D-PBS is used to wash cells during preparation and serial transfer. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells. Used to wash cells during preparation and serial transfer.Specifications
No Calcium, No Magnesium | |
10 X | |
Liquid (as 10X) Inorganic Salts: Potassium Chloride [KCl]: 2000.00 mg/L; Potassium Phosphate Monobasic [KH2PO4]: 2000.00 mg/L; Sodium Chloride [NaCl]: 80000.00 mg/L; Sodium Phosphate Dibasic [Na2HPO4]: 11500.00 mg/L | |
Sterile |
500 mL | |
DPBS | |
6.5 to 7.5 | |
Solution |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.