missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS (10X), pH 7.4
GREENER_CHOICE

PBS (10X), pH 7.4

£20.97 - £189.00

Specifications

Capacity 500 mL
Content And Storage 15°to 30°C
Dilution 10 X
Product Type Phosphate Buffered Saline (PBS)
For Use With (Application) Mammalian Cell Culture
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11504526
GREENER_CHOICE
View Documents
Gibco™
70011051
10 x 500 mL
£189.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11594516
GREENER_CHOICE
View Documents Promotion Details
Gibco™
70011036
500mL
£20.97
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ PBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ PBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ PBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.
Specifications

Specifications

500 mL
10 X
Mammalian Cell Culture
7.3 to 7.5
Liquid
15°to 30°C
Phosphate Buffered Saline (PBS)
2800 to 3100 mOsm/kg
For in-vitro diagnostic use
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 36 Month(s)

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ PBS (10X), pH 7.4

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.