missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBS, 10X Solution, Molecular Biology Grade, Thermo Scientific Chemicals
For biological research. Thermo Scientific™ PBS, 10X Solution, Molecular Biology Grade is 0.2μm filtered and dispensed into durable, square nalgene bottles and upright carton.
£39.62 - £141.44
Specifications
Formulation | 80.6 mM sodium phosphate, 19.4 mM potassium phosphate, 27 mM KCl and 1.37 M NaCl in high purity dH2O |
---|---|
pH | 7.4 |
Form | Solution |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15815418
|
Thermo Scientific
J75889.AE |
100 mL |
£39.62
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15825418
|
Thermo Scientific
J75889.K2 |
1 L |
£75.70
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15835418
|
Thermo Scientific
J75889.K8 |
5 L |
£141.44
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Consists of 80.6mM sodium phosphate, 19.4mM potassium phosphate, 27mM KCl and 1.37M NaCl in high purity dH2O
- Filtered and dispensed into durable, square Nalgene bottles and upright carton with convenient dispensing spout
- Dilute to 1X with high purity dH2O for a solution consisting of 10mM phosphate
Specifications
80.6 mM sodium phosphate, 19.4 mM potassium phosphate, 27 mM KCl and 1.37 M NaCl in high purity dH2O | |
7.4 | |
Solution |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PBS, 10X Solution, Molecular Biology Grade, Thermo Scientific Chemicals