missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBS, Phosphate Buffered Saline, 1X Powder, pH 7.4, Fisher BioReagents
Brand: Fisher BioReagents BP661-50
209.37 GBP valid until 2024-12-31
Use promo code "21951" to get your promotional price.
This item is not returnable.
View return policy
Description
- Provides a convenient way to make 50L of standard 1X phosphate buffer
- Reconstitute contents of bottle in ultrapure water, such as BP2819, to yield a 1X PBS solution containing 0.137M NaCl, 0.0027M KCl, and 0.0119M phosphates
- Tested for the absence of DNase, RNase, and protease
- PBS is a standard buffer used in many biochemical applications to maintain pH
- Intended for laboratory research use only
Specifications
White | |
Poly Bottle | |
Powder |
Room temperature | |
492 grams (makes 50L solution) |
Safety and Handling
Product Identifier
- PBS Phosphate Buffered Saline