missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS, pH 7.4

Product Code. 15390314
Click to view available options
Quantity:
500 mL
5 L
10 L
10 x 500 mL
Unit Size:
10 L
5 L
500mL
Pack of 10
This item is not returnable. View return policy

Product Code. 15390314

Brand: Gibco™ 10010002

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Gibco PBS, pH 7.4, is a balanced salt solution ideal for cell culture applications. It lacks calcium, magnesium, and phenol red, preventing cell adhesion and assay interference. Trusted for quality, Gibco PBS ensures reliable results in cell washing, transport, and reagent preparation.

Gibco Phosphate Buffered Saline (PBS), pH 7.4, is a balanced salt solution widely used in various cell culture applications. This product is specifically formulated without calcium, magnesium, and phenol red, making it ideal for a range of laboratory procedures.

Key features

  • Without calcium and magnesium—the absence of calcium and magnesium prevents unwanted cell adhesion and aggregation, which is crucial during cell dissociation and flow cytometry
  • Without phenol red—the lack of phenol red ensures that there is no interference with optical measurements and prevents potential effects on cell physiology

Applications

  • Washing cells before dissociation—PBS is commonly used to rinse cells before enzymatic dissociation, ensuring that chelators are removed from the culture
  • Transporting cells or tissue—PBS provides a stable environment for cells or tissues during transportation
  • Diluting cells for counting—PBS is used to dilute cell suspensions for accurate counting
  • Preparing reagents—PBS serves as a buffer in the preparation of various reagents

Why choose PBS over DPBS

  • Simplicity—PBS is a simpler formulation compared to Dulbecco's Phosphate-Buffered Saline (DPBS), which includes additional components like potassium chloride and may be formulated with or without calcium, magnesium, glucose, and pyruvate. This simplicity makes PBS a versatile choice for general cell culture applications.
  • Specific applications—PBS is preferred for applications where the presence of potassium or other additives in DPBS might interfere with experimental outcomes

Why choose pH 7.4 over pH 7.2

  • Physiological relevance—pH 7.4 is closer to the physiological pH of human blood, making it more suitable for maintaining the natural state of cells during experiments. This pH is particularly important for applications involving mammalian cells.

Why choose Gibco PBS, pH 7.4

  • Quality and reliability—manufacture of Gibco PBS, pH 7.4, is cGMP-compliant under the ISO 13485 standard, ensuring high standards of quality and consistency
  • Extensive testing—lot release testing of osmolality, pH levels, stability, endotoxins, and sterility ensures the quality of the product
  • Proven track record—Gibco cell culture products have a long-standing reputation in the scientific community, with numerous publications citing the use of Gibco PBS, reflecting its widespread acceptance and trust among researchers

Gibco PBS, pH 7.4, is a reliable and high-quality solution for various applications. Its formulation without calcium, magnesium, and phenol red makes it versatile and suitable for specific experimental needs. Choosing Gibco PBS ensures consistency, quality, and support from a trusted brand in the scientific community.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 1 X
For Use With (Application) Mammalian Cell Culture
pH 7.3 to 7.5
Physical Form Liquid
Product Line Gibco
Quantity 5 L
Solution Type Phosphate Buffered Saline
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 280 - 315 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.