Learn More
Gibco™ PBS, pH 7.4

PBS (phosphate buffered saline) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
£19.48 - £26.95
Specifications
Additive | No Sodium Pyruvate |
---|---|
Classification | Animal Origin-Free |
Color | Clear |
Dilution | 1 X |
Product Line | Gibco™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10728775
![]() |
Gibco™
10010031 |
1000 mL |
£26.95
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15374875
![]() |
Gibco™
10010023 |
500 mL |
£19.48
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PBS (phosphate buffered saline) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. We offer a variety of PBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
This PBS is manufactured as follows:
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Specifications
No Sodium Pyruvate | |
Clear | |
Gibco™ | |
Phosphate Buffered Saline | |
280 - 315 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Animal Origin-Free | |
1 X | |
Phosphate Buffered Saline (PBS) | |
Washing Cells Before Dissociation; Diluting Cells for Counting and Preparing Reagents; Transporting Cells or Tissue | |
7.4 | |
Room Temperature | |
Liquid |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.