Learn More
PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™
Commonly used buffer
Brand: Fisher BioReagents BP399-500
27.65 GBP valid until 2024-12-31
Use promo code "21951" to get your promotional price.
This item is not returnable.
View return policy
Description
Commonly used buffer
- Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X PBS solution - simply dilute as needed
- 10X solution contains 1.37M NaCl, 0.027M KCl and 0.119M phosphates
- pH of a 1X solution made from this concentrated solution is ∼7.4. To prepare a 1X solution, mix one volume of Fisher BioReagents 10X PBS solution with 9 volumes of ultrapure water, such as BP2819
- Filtered and tested for the absence of DNase, RNase, and protease
- PBS is used in many biochemical applications to maintain pH
- Intended for laboratory research use only
Specifications
Room temperature | |
Poly Bottle | |
500mL | |
Liquid |