Learn More
PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™
Commonly used buffer
Brand: Fisherbrand
81.13 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 32
Additional Details : CAS Number : 7778-77-0 Weight : 4.05000kg
Description
Commonly used buffer
- Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X PBS solution - simply dilute as needed
- 10X solution contains 1.37M NaCl, 0.027M KCl and 0.119M phosphates
- pH of a 1X solution made from this concentrated solution is ∼7.4. To prepare a 1X solution, mix one volume of Fisher BioReagents 10X PBS solution with 9 volumes of ultrapure water, such as BP2819
- Filtered and tested for the absence of DNase, RNase, and protease
- PBS is used in many biochemical applications to maintain pH
- Intended for laboratory research use only
Specifications
Room temperature | |
Poly Cube | |
4L | |
Liquid |