missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Phosphate Buffered Saline Tablets (PBS)

Product Code. p-27778 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 Tablets
200 Tablets
500 Tablets
Unit Size:
100 tablets
200 tablets
500 tablets
This item is not returnable. View return policy

Product Code. 11435015

Brand: MP Biomedicals™ 092810305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phosphate Buffered Saline Tablets (PBS) are a balanced salt mixture for easy PBS buffer preparation for a variety of applications, including reagent preparation, diluting cells for flow cytometry, and cell culture reagent.

  • No need for weighing and mixing chemicals
  • Simply dissolve PBS tablets in water
  • One tablet dissolved in 100mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
  • Research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Physical Form Tablet
Quantity 100 Tablets
Molecular Formula ClK
MDL Number MFCD00011360
Includes No Calcium, No Magnesium
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.