missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ PBT Red Screw Caps
Click to view available options
Thread Size (Metric):
GL 14
GL 18
GL 45
Unit Size:
Pack of 10
Specifications
Specifications
| Color | Red |
| For Use With (Application) | Screw thread tubes |
| Includes | DIN thread (GL) |
| Material | PBT |
| Max. Temperature (Metric) | 180°C |
| Quantity | 10Pack |
| Screw Material | PBT |
| Product Line | Cap |
| Thread Size (Metric) | GL 14 |
| Product Type | Screw Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction