missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ PBT Red Screw Caps with Aperture

Brand:  Lenz Laborglasinstrumente™ 01321025

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10692462

  • £27.00 / Pack of 10
Estimated Shipment: 27-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Screw thread tubes
PBT
10Pack
Cap
Screw Cap
GL 25
Screw Cap
DIN thread (GL) and aperture
180°C
PBT
Red
PBT
15 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers