Learn More
Kartell™ PC Safety Box
Made of polycarbonate for strength, chemical resistance, and thermo stability. Kartell™ PC Safety Box is designed to provide a high degree of safety when transporting antiblastic drugs from the preparation center to the administration site.
Brand: Kartell™ 0056900
152.98 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : A0
Additional Details : Weight : 1.35000kg
Description
- Fully autoclavable
- Transport tubes, containers, vials, plastic bags and ready-to-use syringes
- Airtight cover has a hermmetic silicone safety seal to ensure containment of fluids or anti-tumor drugs and protect against leakage and/or accidental spillage from bottles or seepage from plastic bags
- 4 safety locks prevent accidental opening if box is dropped
- Crystal clear to allow inspection of all contents before opening
- Beveled edges and specially designed bottom guarantee safe, simple and efficient cleaning
- Indelible biological hazard symbol molded on outside of cover
- Stainless steel AISI 304 handle for ease of transport
Specifications
Clear | |
CE-IVD | |
330 x 175 mm | |
180 mm | |
175 mm |
Polycarbonate | |
Lid | |
330 x 175 x 180 mm | |
330 mm |