missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDH1 (Protocadherin 1) Monoclonal Antibody (CSTEM31), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: eBioscience 53915741
This item is not returnable.
View return policy
Description
This monoclonal antibody CSTEM31 reacts with human Protocadherin-1, also known as PCDH1 and cadherin-like protein 1. Applications Reported: This CSTEM31 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CSTEM31 antibody has been pre-diluted and tested by flow cytometric analysis of human iPSCs. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
Protocadherin 1, also known as PCDH1 and cadherin-like protein 1, is a transmembrane protein involved in cell-cell adhesion. Protocadherin 1 belongs to a group of cadherin-like proteins termed delta1-protocadherins, which consists of protocadherin-1, -7, -9, and -11(X/Y). The delta1-protocadherins have seven extracellular cadherin-repeats, a transmembrane domain, and a cytoplasmic domain, and can be expressed in two isoforms, short or long. Expression of protocadherin 1 is regulated both temporally and spatially, and plays a role both in early development and mature tissues. In mice, it is expressed in a number of tissues and organs, including the brain, spinal cord, peripheral ganglia, inner ear, hair follicles, kidney, reproductive organs, prostate, and the seminal gland. Notably, protocadherin 1 is a susceptibility gene for Bronchial Hyperresponsiveness in humans. Expression of protocadherin 1 has also been identified on human pluripotent stem cells.Specifications
PCDH1 (Protocadherin 1) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q08174 | |
PCDH1 | |
Affinity chromatography | |
RUO | |
5097 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
CSTEM31 | |
Alexa Fluor 488 | |
PCDH1 | |
2010005A06Rik; AI585920; cadherin-like 1; cadherin-like protein 1; PC42; PCDH1; PCDH42; protocadherin 1; protocadherin 1 (cadherin-like 1); protocadherin 42; protocadherin-1; Protocadherin-42 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b κ |