missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress PCI-33380

MedChemExpress PCI-33380 is an irreversible and selective Bruton's Tyrosine Kinase (BTK) inhibitor (fluorescent probe).

Encompass_Preferred

£421.08 - £2283.27

Specifications

Product Type Reference compound
Molecular Weight (g/mol) 855.78 g/mol
Solubility DMSO : ≥ 50 mg/mL (58.43 mM)
Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16499516
Encompass_Preferred
View Documents
MedChemExpress
HY-100335
50 mg
£2283.27
50mg
View alternative products
Save Up to 
»
Estimated Shipment: 27-03-2025
to see stock.
Add to basket
 
16495687
Encompass_Preferred
View Documents
MedChemExpress
HY-100335
5 mg
£421.08
5mg
View alternative products
Save Up to 
»
Estimated Shipment: 27-03-2025
to see stock.
Add to basket
 
16472948
Encompass_Preferred
View Documents
MedChemExpress
HY-100335
10 mg
£660.66
10mg
View alternative products
Save Up to 
»
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
 
16412556
Encompass_Preferred
View Documents
MedChemExpress
HY-100335
25 mg
£1442.32
25mg
View alternative products
Save Up to 
»
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Reference compound
DMSO : ≥ 50 mg/mL (58.43 mM)
Room temperature
855.78 g/mol
Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
1022899-36-0
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
MedChemExpress PCI-33380

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.