Learn More
BRAND™ PCR 8-Strip Caps
Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples.PCR 8-Strip Caps are autoclavable and RNase-, DNase-, and DNA-free. Domed caps are available in a variety of colors for sample identification.
Brand:  BRAND™ 781342
27.10 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10633493
  • £32.10 / Pack of 125
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.
  • Available flat or domed
  • The domed caps are available in a variety of colors for sample identification
  • Autoclavable
  • RNase-, DNase-, and DNA-free.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yellow
Domed
Polypropylene
Yes
Autoclavable
Single-use
Microplates
Non-sterile
Yes
8-strip Domed Cap
Yes
Cap Strip