missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PCR 8 x 8 Rotor with Screw-On-Lid for Sorvall Legend Micro, MicroCL, Pico and Fresco Centrifuges

Increase throughput and accelerate research. This Thermo Scientific™ PCR 8 x 8 Rotor with Screw-On-Lid accommodates eight eight-place PCR strips or 64 individual PCR tubes.

Brand:  Thermo Scientific™ 75003489

Product Code. 10611045

  • £716.21 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Accommodate four eight-place PCR strips or 32 individual PCR tubes with Thermo Scientific™ PCR 4 x 8 Rotor with Screw-On Lid.

Specifications:
  • Capacity: 64 × 0.2mL
  • Max. Speed/RCF: 14,800rpm/17,100 x g

Ordering Information:

Replacement lids sold separately; order rotor body (75003489) and components to complete assembly. For replacement lids order 75003406. Contact your Sales Representative for information.

Compatible with:

Thermo Scientific MicroCL 21/21R Microcentrifuge Series.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PCR 8 x 8 Rotor with Screw-On Lid
Rotor
17,100 x g
7.0 cm
6 mm
60°
Engineered Polymer
64 x 0.2 mL
14,800 rpm
4.4 cm
Sorvall Legend Micro 17/17R/21/21R, Pico and Fresco 17/21, MicroCL 17/17R/21/21R Centrifuges
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.