Learn More
Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes
Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.
Brand: Eppendorf™ 0030123301
47.79 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 96
Additional Details : Weight : 0.32400kg
Description
Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.
- Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
- g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Precise lid sealing for minimal evaporation rates during long-term storage
- Guaranteed functionality from -86 to 100°C
- Large frosted lid and surface on the side for easy labeling
- Autoclavable when open (121°C, 20 min)
Specifications
Natural | |
500 | |
Autoclavable | |
Centrifuge Tube |
0.5 mL | |
Polypropylene | |
Yes |