missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ PCR-Cooler

Product Code. 10275091 Shop All Eppendorf Products
Click to view available options
Color:
Blue
Blue, Pink
Pink
Unit Size:
1 set
Each
This item is not returnable. View return policy

Product Code. 10275091

Brand: Eppendorf™ 3881000023

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Innovative solution for protecting, transporting and storing sensitive samples as well as for safely stopping reactions.

  • Maintains a sample temperature of 0°C for over an hour and then changes color when the temperature has exceeded 7°C
  • Superior to an ice bath, it is easy to handle, space saving and free of contamination, which makes it the perfect preparation for PCR reactions
  • Temperature of single tubes is indicated by the color of the individual bores
  • Samples are prevented from freezing by the specially developed cone contour
  • Cooling aggregate in the universal 96 well format for 0.2mL and 0.5mL PCR tubes, for tube strips, plates and individual tubes
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) PCR Reaction Setup
Type PCR Cooler
Color Pink
Capacity (Metric) 0.2 mL
Holds 96
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.