missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PCR™ Cooler, 96-Well, 2 Pack
The PCR™ Cooler is ideal for thermosensitive applications, PCR, or cell-based assays, as well as gentle sample thawing and stopping of reactions. The unit also provides a dependable cooling station for pipetting and sample storage. Perfect for transporting samples and reactions between preparation, thermal cycling and post-PCR procedure areas.
£132.00 - £137.77
Specifications
Thermal Accuracy | ±1°C |
---|---|
Temperature Range (Metric) | 0°C to 5°C |
Content And Storage | Lid, Rack |
For Use With (Equipment) | Shakers, Mixers, Vortexers |
Capacity | Tubes |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
16300412
|
Fisherbrand™
120727F |
Purple and Pink |
£137.77
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16310412
|
Fisherbrand™
120728F |
Dark Blue and Light Blue |
£136.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16320412
|
Fisherbrand™
120729F |
Purple and Pink and Dark Blue and Light Blue |
£132.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Modern patented design cools faster in freezers and stays cold longer on the bench top
- PCR™ Cooler maintains a sample temperature of 0 °C (with 2 hours pre-cooling at -20 °C) for over an hour
- When the temperature ceiling of 6 °C has been exceeded, users are signaled by a contrasting color change of the cooler, or even a single well
- Cools industry standard format 0.2 mL and 0.5 mL PCR™ tubes, strips and single format, as well as most 96 well plates, in both standard and low profile well shape
- Better alternative then ice baths as dry incubation reduces the risk of contamination
- Freeze cooler in an upright position enabling cold storage of tubes
- Well design prohibits the freezing of samples
- Fully sealed and sonic welded cooler keeps coolant contained offering worry free usage
- Save space with compact footprint and stacking ability
- The lid is ambidextrous; it can be switched to accommodate left or right-hand orientation and is detachable
- Robust cooler made from engineered polypropylene and the lid is made from polycarbonate
- US Patent Pending, Community Design
- No: 004101756-0001-0002
- Two-Year Warranty
Specifications
±1°C | |
Lid, Rack | |
Tubes | |
14.8 cm | |
PCR Applications | |
2 | |
.27 kg (0.6 lb.) |
0°C to 5°C | |
Shakers, Mixers, Vortexers | |
1.9 cm | |
4.8 cm | |
Fisherbrand™ PCR™ Cooler, 96-Well, 2 Pack | |
PCR Rack |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ PCR™ Cooler, 96-Well, 2 Pack